Learn More
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chair - Desk Height with ESD Casters in ESD Vinyl
Shop All Fisherbrand ProductsDescription
- Adjustable from 18.5-23.75 inches from the top of the seat to the floor
- Features manual back height and back depth adjustments
- Electro-static dissipating components to ensure proper grounding
- Cast-aluminum base
- Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
- Electro-static dissipating vinyl
- Class 10/ISO 4 Clean Room
- Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
| Product Type | Cleanroom ESD Chair |
| With Lumbar Support | Yes |
| Autoclavable | Not Autoclavable |
| Base Material | Polished Cast Aluminum |
| Adjustable | Yes |
| Certifications/Compliance | Class 10/ISO Class 4 |
| Load Bearing Capacity (Metric) | 136 kg |
| Back Style | Standard |
| Adjustable (Height) | 18.5 to 23.75 in. |
| Seat Material | ESD Upholstery |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.